Product Details

SNP ID
rs183487896
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:41690609 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCCTCTAGCTCCCAACACCATCT[A/G]CCTGATCTAAGCTTTGCCATGTTGT
Phenotype
MIM: 608797 MIM: 601304
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C22orf46 PubMed Links

Gene Details

Gene
C22orf46
Gene Name
chromosome 22 open reading frame 46
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142964.1 67 UTR 5 NP_001136436.1
XM_011530373.2 67 UTR 5 XP_011528675.1
XM_017028939.1 67 Intron XP_016884428.1
Gene
MEI1
Gene Name
meiotic double-stranded break formation protein 1
There are no transcripts associated with this gene.

Gene
SNU13
Gene Name
SNU13 homolog, small nuclear ribonucleoprotein (U4/U6.U5)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003796.1 67 Intron NP_001003796.1
NM_005008.3 67 Intron NP_004999.1
XM_017028811.1 67 Intron XP_016884300.1

View Full Product Details