Product Details

SNP ID
hCV27852194
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:78752052 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTAATCACTGAGAACAGGATTTT[A/G]TCAAATTGAGGCCCATATCAGTTTC
Phenotype
MIM: 300086
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LPAR4 PubMed Links

Gene Details

Gene
LPAR4
Gene Name
lysophosphatidic acid receptor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278000.1 Intron NP_001264929.1
NM_005296.2 Intron NP_005287.1

View Full Product Details