Product Details

SNP ID
rs7111860
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:26994143 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCCAGTCCAGAAATTGAAATACTA[G/T]CAGGGGGCAAGAGCCTTTCTCTCCA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FIBIN PubMed Links

Gene Details

Gene
FIBIN
Gene Name
fin bud initiation factor homolog (zebrafish)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203371.1 63 UTR 5 NP_976249.1

View Full Product Details