Product Details

SNP ID
rs565262308
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:35051519 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGTGAGATGAGAATCCTTAGAGT[C/T]TGGTTCACTATGACTGGGATTGTTC
Phenotype
MIM: 612471 MIM: 176761
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AGXT2 PubMed Links

Gene Details

Gene
AGXT2
Gene Name
alanine--glyoxylate aminotransferase 2
There are no transcripts associated with this gene.

Gene
PRLR
Gene Name
prolactin receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000949.6 Intron NP_000940.1
NM_001204314.2 Intron NP_001191243.1
NM_001204315.1 Intron NP_001191244.1
NM_001204316.1 Intron NP_001191245.1
NM_001204317.1 Intron NP_001191246.1
NM_001204318.1 Intron NP_001191247.1
XM_006714484.2 Intron XP_006714547.1
XM_011514068.2 Intron XP_011512370.1
XM_011514069.2 Intron XP_011512371.1
XM_017009645.1 Intron XP_016865134.1
XM_017009646.1 Intron XP_016865135.1

View Full Product Details