Product Details

SNP ID
rs7882094
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:100969664 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACCGTGACAACCATTATTAGGACA[C/T]TGCCATAGCTTTTATAAATGGACAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARL13A PubMed Links

Gene Details

Gene
ARL13A
Gene Name
ADP ribosylation factor like GTPase 13A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001162491.1 654 Intron NP_001155963.1
XM_011530950.2 654 UTR 5 XP_011529252.1
XM_011530951.2 654 UTR 5 XP_011529253.1
Gene
TRMT2B
Gene Name
tRNA methyltransferase 2 homolog B
There are no transcripts associated with this gene.

View Full Product Details