Product Details

SNP ID
rs148066688
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:30970574 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATAAGCCTATTTTATCCTTTGGC[C/T]CGTCAACTCTGTTATCTGCTGCTTG
Phenotype
MIM: 616661 MIM: 614971
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MORC2 PubMed Links

Gene Details

Gene
MORC2
Gene Name
MORC family CW-type zinc finger 2
There are no transcripts associated with this gene.

Gene
TUG1
Gene Name
taurine up-regulated 1 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details