Product Details

SNP ID
rs540081043
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:68638137 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTATTTGTGCTTTTAAAATTGTGG[C/T]CATAATTTATCATCATTACTTTTCA
Phenotype
MIM: 602684
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADGRB3 PubMed Links

Gene Details

Gene
ADGRB3
Gene Name
adhesion G protein-coupled receptor B3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001704.2 Intron NP_001695.1
XM_005248752.2 Intron XP_005248809.1
XM_006715534.3 Intron XP_006715597.1
XM_011536009.2 Intron XP_011534311.1
XM_011536010.2 Intron XP_011534312.1
XM_011536011.2 Intron XP_011534313.1
XM_011536012.2 Intron XP_011534314.1
XM_017011140.1 Intron XP_016866629.1
XM_017011141.1 Intron XP_016866630.1
XM_017011142.1 Intron XP_016866631.1
XM_017011143.1 Intron XP_016866632.1
XM_017011144.1 Intron XP_016866633.1
Gene
LOC101928307
Gene Name
uncharacterized LOC101928307
There are no transcripts associated with this gene.

View Full Product Details