Product Details

SNP ID
rs115760234
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:39322529 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTTTTCTACCTTTTTGGTTTCAC[A/G]ACTTGAGCATGTTTTTATTATTTTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADAM5 PubMed Links

Gene Details

Gene
ADAM5
Gene Name
ADAM metallopeptidase domain 5 (pseudogene)
There are no transcripts associated with this gene.

View Full Product Details