Product Details

SNP ID
rs41275303
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:160010396 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGAATTCCAGTGAAAGGAAATTA[C/T]GGTGTGTTTTTTGGCTGTAATGCAG
Phenotype
MIM: 601963
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TTC1 PubMed Links

Gene Details

Gene
TTC1
Gene Name
tetratricopeptide repeat domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282500.1 Intron NP_001269429.1
NM_003314.2 Intron NP_003305.1

View Full Product Details