Product Details

SNP ID
rs78007930
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:73497745 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGAATGTTATGTTGGCAGCTCCGC[A/G]TTCTAGGCCTAAGGTTTGGAGGGCA
Phenotype
MIM: 114106
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PPP3CB PubMed Links

Gene Details

Gene
PPP3CB
Gene Name
protein phosphatase 3 catalytic subunit beta
There are no transcripts associated with this gene.

Gene
PPP3CB-AS1
Gene Name
PPP3CB antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

Gene
USP54
Gene Name
ubiquitin specific peptidase 54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320437.1 5843 UTR 3 NP_001307366.1
NM_001320441.1 5843 Intron NP_001307370.1
NM_152586.3 5843 UTR 3 NP_689799.3
XM_005269582.2 5843 UTR 3 XP_005269639.1
XM_011539368.2 5843 UTR 3 XP_011537670.1
XM_017015759.1 5843 UTR 3 XP_016871248.1
XM_017015760.1 5843 UTR 3 XP_016871249.1
XM_017015761.1 5843 UTR 3 XP_016871250.1
XM_017015762.1 5843 UTR 3 XP_016871251.1
XM_017015763.1 5843 UTR 3 XP_016871252.1
XM_017015764.1 5843 UTR 3 XP_016871253.1
XM_017015765.1 5843 UTR 3 XP_016871254.1
XM_017015766.1 5843 UTR 3 XP_016871255.1
XM_017015767.1 5843 UTR 3 XP_016871256.1
XM_017015768.1 5843 UTR 3 XP_016871257.1
XM_017015769.1 5843 UTR 3 XP_016871258.1
XM_017015770.1 5843 UTR 3 XP_016871259.1
XM_017015771.1 5843 UTR 3 XP_016871260.1
XM_017015772.1 5843 Intron XP_016871261.1
XM_017015773.1 5843 UTR 3 XP_016871262.1
XM_017015774.1 5843 UTR 3 XP_016871263.1
XM_017015775.1 5843 UTR 3 XP_016871264.1
XM_017015776.1 5843 UTR 3 XP_016871265.1
XM_017015777.1 5843 UTR 3 XP_016871266.1
XM_017015778.1 5843 UTR 3 XP_016871267.1
XM_017015779.1 5843 UTR 3 XP_016871268.1
XM_017015780.1 5843 Intron XP_016871269.1
XM_017015781.1 5843 UTR 3 XP_016871270.1
XM_017015782.1 5843 UTR 3 XP_016871271.1
XM_017015783.1 5843 UTR 3 XP_016871272.1
XM_017015784.1 5843 UTR 3 XP_016871273.1
XM_017015785.1 5843 UTR 3 XP_016871274.1
XM_017015786.1 5843 UTR 3 XP_016871275.1

View Full Product Details