Product Details

SNP ID
rs371934738
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:50476598 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGCTCCTGCTCTTGCTGATGGC[C/G]GTAGCAGCGCCCAGTCGAGCCCGGG
Phenotype
MIM: 614545
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
EMC10 PubMed Links

Gene Details

Gene
EMC10
Gene Name
ER membrane protein complex subunit 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175063.5 149 Silent Mutation GCC,GCG A18A NP_778233.4
NM_206538.3 149 Silent Mutation GCC,GCG A18A NP_996261.1
XM_006723163.3 149 Silent Mutation GCC,GCG A18A XP_006723226.1
XM_011526818.2 149 Silent Mutation GCC,GCG A18A XP_011525120.1
Gene
FAM71E1
Gene Name
family with sequence similarity 71 member E1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308429.1 149 UTR 5 NP_001295358.1
NM_138411.2 149 UTR 5 NP_612420.1
XM_011526405.2 149 Intron XP_011524707.2
XM_011526406.2 149 Intron XP_011524708.2

View Full Product Details