Product Details

SNP ID
rs181390772
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:154222539 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTGTTTTGTTTTTCATCTTTAAA[A/G]CTCCTCCTTGCTATGTCCTAACTGG
Phenotype
MIM: 616472
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1orf43 PubMed Links

Gene Details

Gene
C1orf43
Gene Name
chromosome 1 open reading frame 43
There are no transcripts associated with this gene.

Gene
UBAP2L
Gene Name
ubiquitin associated protein 2 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127320.2 Intron NP_001120792.1
NM_001287815.1 Intron NP_001274744.1
NM_001287816.1 Intron NP_001274745.1
NM_014847.3 Intron NP_055662.3
XM_005245658.3 Intron XP_005245715.1
XM_005245667.3 Intron XP_005245724.1
XM_005245668.3 Intron XP_005245725.1
XM_005245669.3 Intron XP_005245726.1
XM_005245670.3 Intron XP_005245727.1
XM_005245672.3 Intron XP_005245729.1
XM_005245673.3 Intron XP_005245730.1
XM_005245674.1 Intron XP_005245731.1
XM_011510215.2 Intron XP_011508517.1
XM_011510216.1 Intron XP_011508518.1
XM_017002972.1 Intron XP_016858461.1
XM_017002973.1 Intron XP_016858462.1
XM_017002974.1 Intron XP_016858463.1
XM_017002975.1 Intron XP_016858464.1
XM_017002976.1 Intron XP_016858465.1
XM_017002977.1 Intron XP_016858466.1
XM_017002978.1 Intron XP_016858467.1
XM_017002979.1 Intron XP_016858468.1
XM_017002980.1 Intron XP_016858469.1
XM_017002981.1 Intron XP_016858470.1
XM_017002982.1 Intron XP_016858471.1
XM_017002983.1 Intron XP_016858472.1
XM_017002984.1 Intron XP_016858473.1
XM_017002985.1 Intron XP_016858474.1
XM_017002986.1 Intron XP_016858475.1
XM_017002987.1 Intron XP_016858476.1
XM_017002988.1 Intron XP_016858477.1
XM_017002989.1 Intron XP_016858478.1
XM_017002990.1 Intron XP_016858479.1
XM_017002991.1 Intron XP_016858480.1

View Full Product Details