Product Details

SNP ID
rs11178998
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:71938935 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCTCAATCTCCGCCAGCGCTGCT[A/G]CTGCCCCTCTAGTACCCCCTGCTGC
Phenotype
MIM: 607478
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
TPH2 PubMed Links

Gene Details

Gene
TPH2
Gene Name
tryptophan hydroxylase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173353.3 90 UTR 5 NP_775489.2

View Full Product Details