Product Details
- SNP ID
-
hCV27855866
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:106308385 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGACTCCTGACTCCCAATTATTTTA[A/G]TCTTTGCACTTTTCTGCCTTGTTAA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TCP11L2
PubMed Links
Gene Details
- Gene
- TCP11L2
- Gene Name
- t-complex 11 like 2
View Full Product Details