Product Details

SNP ID
rs12044983
Assay Type
Functionally tested
NCBI dbSNP Submissions
28
Location
Chr.1:91642562 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGAGCCCATGGAGGTAGAAGAAGC[A/G]GCAAAAGAAGACAAAGAAGAATCTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HSP90B3P PubMed Links

Gene Details

Gene
HSP90B3P
Gene Name
heat shock protein 90 beta family member 3, pseudogene
There are no transcripts associated with this gene.

View Full Product Details