Product Details

SNP ID
rs11098947
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:128074596 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTAAGAACTCTTAGGTTATTATAA[A/G]TGTATTTTTCTTCTAATTGTATTAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LARP1B PubMed Links

Gene Details

Gene
LARP1B
Gene Name
La ribonucleoprotein domain family member 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278604.1 Intron NP_001265533.1
NM_018078.3 Intron NP_060548.2
NM_032239.3 Intron NP_115615.2
NM_178043.2 Intron NP_835144.1
XM_011532057.2 Intron XP_011530359.2
XM_011532058.2 Intron XP_011530360.2
XM_011532059.2 Intron XP_011530361.2
XM_011532060.2 Intron XP_011530362.2
XM_011532061.2 Intron XP_011530363.1
XM_011532062.2 Intron XP_011530364.2
XM_011532063.2 Intron XP_011530365.2
XM_011532064.2 Intron XP_011530366.2
XM_011532065.2 Intron XP_011530367.1
XM_011532067.1 Intron XP_011530369.1
XM_011532068.1 Intron XP_011530370.1
XM_011532069.1 Intron XP_011530371.1
XM_011532070.2 Intron XP_011530372.2
XM_011532072.2 Intron XP_011530374.2
XM_011532073.2 Intron XP_011530375.2
XM_011532075.1 Intron XP_011530377.1
XM_011532076.2 Intron XP_011530378.1
XM_011532077.2 Intron XP_011530379.1
XM_017008338.1 Intron XP_016863827.1
XM_017008339.1 Intron XP_016863828.1
XM_017008340.1 Intron XP_016863829.1
XM_017008341.1 Intron XP_016863830.1
XM_017008342.1 Intron XP_016863831.1
XM_017008343.1 Intron XP_016863832.1
XM_017008344.1 Intron XP_016863833.1
XM_017008345.1 Intron XP_016863834.1
XM_017008346.1 Intron XP_016863835.1
XM_017008347.1 Intron XP_016863836.1
XM_017008348.1 Intron XP_016863837.1
XM_017008349.1 Intron XP_016863838.1
XM_017008350.1 Intron XP_016863839.1
XM_017008351.1 Intron XP_016863840.1
XM_017008352.1 Intron XP_016863841.1
XM_017008353.1 Intron XP_016863842.1

View Full Product Details