Product Details
- SNP ID
-
rs62624977
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:37761964 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGAAATTGGAAGGGAGTACTCAGAA[A/C]GTAGAGTCACCTGTGCTGCAGGGGC
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TRMT10B
PubMed Links
Gene Details
- Gene
- TRMT10B
- Gene Name
- tRNA methyltransferase 10B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001286950.1 |
139 |
Missense Mutation |
AAA,AAC |
K11N |
NP_001273879.1 |
NM_001286951.1 |
139 |
UTR 5 |
|
|
NP_001273880.1 |
NM_001286952.1 |
139 |
Missense Mutation |
AAA,AAC |
K11N |
NP_001273881.1 |
NM_001286953.1 |
139 |
UTR 5 |
|
|
NP_001273882.1 |
NM_001286954.1 |
139 |
UTR 5 |
|
|
NP_001273883.1 |
NM_144964.3 |
139 |
Missense Mutation |
AAA,AAC |
K11N |
NP_659401.2 |
XM_005251375.2 |
139 |
Missense Mutation |
AAA,AAC |
K11N |
XP_005251432.1 |
XM_005251379.2 |
139 |
UTR 5 |
|
|
XP_005251436.1 |
XM_011517735.2 |
139 |
Missense Mutation |
AAA,AAC |
K24N |
XP_011516037.1 |
XM_011517736.2 |
139 |
Missense Mutation |
AAA,AAC |
K24N |
XP_011516038.1 |
XM_011517738.2 |
139 |
Missense Mutation |
AAA,AAC |
K24N |
XP_011516040.1 |
XM_011517739.2 |
139 |
Missense Mutation |
AAA,AAC |
K24N |
XP_011516041.1 |
XM_011517742.1 |
139 |
Missense Mutation |
AAA,AAC |
K24N |
XP_011516044.1 |
XM_011517743.2 |
139 |
Missense Mutation |
AAA,AAC |
K24N |
XP_011516045.1 |
XM_017014313.1 |
139 |
Missense Mutation |
AAA,AAC |
K24N |
XP_016869802.1 |
View Full Product Details