Product Details

SNP ID
hCV27856951
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:122945420 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCCCTCCTCTTTCCAAATCAACC[A/G]TAATATTCTGAGGTACTCAGTGAGA
Phenotype
MIM: 615882
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RABGAP1 PubMed Links

Gene Details

Gene
RABGAP1
Gene Name
RAB GTPase activating protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012197.3 Intron NP_036329.3

View Full Product Details