Product Details

SNP ID
hCV27857227
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:15686061 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTCTATTTACTAATCTCAGTTG[A/C]AGTGGTTTGCCTTACAGAAATAGGC
Phenotype
MIM: 600387 MIM: 605655
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
BST1 PubMed Links

Gene Details

Gene
BST1
Gene Name
bone marrow stromal cell antigen 1
There are no transcripts associated with this gene.

Gene
FAM200B
Gene Name
family with sequence similarity 200 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145191.1 Intron NP_001138663.1
Gene
FBXL5
Gene Name
F-box and leucine rich repeat protein 5
There are no transcripts associated with this gene.

View Full Product Details