Product Details

SNP ID
rs5912637
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:78751941 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGCACCAGACCACATTATTTCCCA[A/C]AGATTGGACTGTGTTATCAAGTGAA
Phenotype
MIM: 300086
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LPAR4 PubMed Links

Gene Details

Gene
LPAR4
Gene Name
lysophosphatidic acid receptor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278000.1 Intron NP_001264929.1
NM_005296.2 Intron NP_005287.1
XM_005262126.3 Intron XP_005262183.1
XM_006724639.3 Intron XP_006724702.1
XM_017029437.1 Intron XP_016884926.1
XM_017029438.1 Intron XP_016884927.1

View Full Product Details