Product Details

SNP ID
rs144941686
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:38399427 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATTTGGGGGAGGCACCCCAACAT[A/T]TATCTTCCTGATCTTGTTTCTAAGC
Phenotype
MIM: 607320 MIM: 609293
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FAM98C PubMed Links

Gene Details

Gene
FAM98C
Gene Name
family with sequence similarity 98 member C
There are no transcripts associated with this gene.

Gene
RASGRP4
Gene Name
RAS guanyl releasing protein 4
There are no transcripts associated with this gene.

Gene
SPRED3
Gene Name
sprouty related EVH1 domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042522.2 4618 UTR 3 NP_001035987.1
XM_006723219.3 4618 UTR 3 XP_006723282.1
XM_011526978.2 4618 UTR 3 XP_011525280.1

View Full Product Details