Product Details

SNP ID
rs73479087
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:60088487 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTTCTGTAAATCCTGCTAGTCTC[A/G]GGCAAAATTATGCTCCAGGAGTCTC
Phenotype
MIM: 147138
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MS4A2 PubMed Links

Gene Details

Gene
MS4A2
Gene Name
membrane spanning 4-domains A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000139.4 227 Intron NP_000130.1
NM_001256916.1 227 Intron NP_001243845.1
XM_005273846.4 227 UTR 5 XP_005273903.1
XM_011544850.2 227 Intron XP_011543152.1
XM_017017362.1 227 UTR 5 XP_016872851.1

View Full Product Details