Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_015041.2 | 193 | Intron | NP_055856.1 | ||
NM_024793.2 | 193 | Intron | NP_079069.1 | ||
XM_006720867.3 | 193 | Intron | XP_006720930.1 | ||
XM_017023068.1 | 193 | Intron | XP_016878557.1 | ||
XM_017023069.1 | 193 | Intron | XP_016878558.1 | ||
XM_017023070.1 | 193 | Intron | XP_016878559.1 | ||
XM_017023071.1 | 193 | UTR 5 | XP_016878560.1 | ||
XM_017023072.1 | 193 | UTR 5 | XP_016878561.1 | ||
XM_017023073.1 | 193 | Intron | XP_016878562.1 |