Product Details

SNP ID
rs28942098
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:193122203 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCGGAGGCGAGGGGGGGGAAGAT[A/G]GCGGACGTGCTTAGCGTCCTGCGAC
Phenotype
MIM: 607393
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDC73 PubMed Links

Gene Details

Gene
CDC73
Gene Name
cell division cycle 73
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024529.4 246 Missense Mutation ATA,ATG I1M NP_078805.3
XM_006711537.3 246 Missense Mutation ATA,ATG I1M XP_006711600.1

View Full Product Details