Product Details

SNP ID
rs121908108
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:31820357 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGAGGGCAGTGCTGGGCCCCCGG[A/C]AGCCCTGCCCCAGCAGACTGCGACA
Phenotype
MIM: 606566
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MYLK2 PubMed Links

Gene Details

Gene
MYLK2
Gene Name
myosin light chain kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033118.3 386 Missense Mutation GAA,GCA E95A NP_149109.1

View Full Product Details