Product Details

SNP ID
rs62637020
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:41473545 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGTGGGGGCCTGCGCCCGCGCTT[C/G]TCCCGCCGCCTGCGCCTGCAGCACC
Phenotype
MIM: 300278
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NYX PubMed Links

Gene Details

Gene
NYX
Gene Name
nyctalopin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022567.2 190 Missense Mutation TCT,TGT S31C NP_072089.1
XM_017029709.1 190 Missense Mutation TCT,TGT S31C XP_016885198.1

View Full Product Details