Product Details

SNP ID
hCV27860520
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:17022626 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACGTACCTTAGGACAGGTCCTTGT[G/T]CAGTTCATGATGGTGTGGCAGCGGT
Phenotype
MIM: 185470
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SDHB PubMed Links

Gene Details

Gene
SDHB
Gene Name
succinate dehydrogenase complex iron sulfur subunit B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003000.2 898 Nonsense Mutation TGA,TGC *249C NP_002991.2

View Full Product Details