Product Details

SNP ID
hCV27860521
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:25809095 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGAAGACCCGCTTTGCCCCTCAG[A/G]GAGCTGTGGCCTGCCTGACTGCCAT
Phenotype
MIM: 606210
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SEPN1 PubMed Links

Gene Details

Gene
SEPN1
Gene Name
selenoprotein N, 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020451.2 872 Missense Mutation AGA,GGA R273G NP_065184.2
NM_206926.1 872 Missense Mutation AGA,GGA R239G NP_996809.1

View Full Product Details