Product Details

SNP ID
rs56059937
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.2:233760534 on Build GRCh38
Set Membership
DME Validated Inventoried
Context Sequence [VIC/FAM]
CACCTTGAAGACGTACCCTGTGCCA[C/T]TCCAAAGGGAGGATGTGAAAGAGTC
Phenotype
MIM: 191740 MIM: 606435 MIM: 606428 MIM: 606429 MIM: 606430 MIM: 606431 MIM: 606432 MIM: 606433 MIM: 606434
Polymorphism
C/T, Transition substitution
Allele Nomenclature
UGT1A1*62,c.247T>C UGT1A1*62,g.247T>C
Literature Links
LOC100286922 PubMed Links

Gene Details

Gene
LOC100286922
Gene Name
DnaJ heat shock protein family (Hsp40) member B3 pseudogene
There are no transcripts associated with this gene.

Gene
UGT1A1
Gene Name
UDP glucuronosyltransferase family 1 member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000463.2 262 Missense Mutation CTC,TTC L83F NP_000454.1
Gene
UGT1A10
Gene Name
UDP glucuronosyltransferase family 1 member A10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019075.2 262 Intron NP_061948.1
Gene
UGT1A3
Gene Name
UDP glucuronosyltransferase family 1 member A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019093.2 262 Intron NP_061966.1
Gene
UGT1A4
Gene Name
UDP glucuronosyltransferase family 1 member A4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007120.2 262 Intron NP_009051.1
Gene
UGT1A5
Gene Name
UDP glucuronosyltransferase family 1 member A5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019078.1 262 Intron NP_061951.1
Gene
UGT1A6
Gene Name
UDP glucuronosyltransferase family 1 member A6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001072.3 262 Intron NP_001063.2
NM_205862.1 262 Intron NP_995584.1
Gene
UGT1A7
Gene Name
UDP glucuronosyltransferase family 1 member A7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019077.2 262 Intron NP_061950.2
Gene
UGT1A8
Gene Name
UDP glucuronosyltransferase family 1 member A8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019076.4 262 Intron NP_061949.3
Gene
UGT1A9
Gene Name
UDP glucuronosyltransferase family 1 member A9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021027.2 262 Intron NP_066307.1

View Full Product Details