Product Details

SNP ID
rs72554627
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.8:142912546 on Build GRCh38
Set Membership
DME Validated Inventoried
Context Sequence [VIC/FAM]
CCTGCTTACGTGGTGCAGCAGCAGC[A/G]GCATCTCTGCCTCTGCCAGGCGCCG
Phenotype
MIM: 124080
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CYP11B2 PubMed Links

Gene Details

Gene
CYP11B2
Gene Name
cytochrome P450 family 11 subfamily B member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000498.3 1385 Missense Mutation CCG,CTG P461L NP_000489.3

View Full Product Details