Product Details

SNP ID
rs121912986
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:87610957 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTTGGGCAGTAGCATGGGCCATT[A/C]CAGTAAGTATGCCTTTCTTAGAAAA
Phenotype
MIM: 125485
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DSPP PubMed Links

Gene Details

Gene
DSPP
Gene Name
dentin sialophosphoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014208.3 Intron NP_055023.2

View Full Product Details