Product Details

SNP ID
rs28933376
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:70598417 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGAAGAGCTTCACATAGGCATCC[A/G]TGGCAGTGAACCAGTCCCCCCACAG
Phenotype
MIM: 170280
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
PRF1 PubMed Links

Gene Details

Gene
PRF1
Gene Name
perforin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083116.1 1465 Missense Mutation ACG,ATG T435M NP_001076585.1
NM_005041.4 1465 Missense Mutation ACG,ATG T435M NP_005032.2

View Full Product Details