Product Details

SNP ID
rs4759190
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55725460 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCAAGCATCACTCTAAGACAAACTC[A/G]GACCATCTCTTTTCGGTCTGAAAAA
Phenotype
MIM: 601444 MIM: 155740 MIM: 600536 MIM: 601617
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BLOC1S1 PubMed Links

Gene Details

Gene
BLOC1S1
Gene Name
biogenesis of lysosomal organelles complex 1 subunit 1
There are no transcripts associated with this gene.

Gene
BLOC1S1-RDH5
Gene Name
BLOC1S1-RDH5 readthrough
There are no transcripts associated with this gene.

Gene
CD63
Gene Name
CD63 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001257389.1 959 UTR 3 NP_001244318.1
NM_001257390.1 959 UTR 3 NP_001244319.1
NM_001257391.1 959 UTR 3 NP_001244320.1
NM_001257392.1 959 UTR 3 NP_001244321.1
NM_001257400.1 959 UTR 3 NP_001244329.1
NM_001257401.1 959 UTR 3 NP_001244330.1
NM_001267698.1 959 UTR 3 NP_001254627.1
NM_001780.5 959 UTR 3 NP_001771.1
Gene
ITGA7
Gene Name
integrin subunit alpha 7
There are no transcripts associated with this gene.

Gene
RDH5
Gene Name
retinol dehydrogenase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199771.1 959 Intron NP_001186700.1
NM_002905.3 959 Intron NP_002896.2

View Full Product Details