Product Details

SNP ID
rs4818989
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:44936901 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCTACAGGGAAGCACGTCATAGCC[A/G]ACAGAATTCTGTACAGACATTAAAG
Phenotype
MIM: 600065
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM207A PubMed Links

Gene Details

Gene
FAM207A
Gene Name
family with sequence similarity 207 member A
There are no transcripts associated with this gene.

Gene
ITGB2
Gene Name
integrin subunit beta 2
There are no transcripts associated with this gene.

Gene
ITGB2-AS1
Gene Name
ITGB2 antisense RNA 1
There are no transcripts associated with this gene.

Gene
LINC01547
Gene Name
long intergenic non-protein coding RNA 1547
There are no transcripts associated with this gene.

View Full Product Details