Product Details

SNP ID
rs4523976
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:6854656 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAGCAATTAGGACCAAGGGAAAG[A/C]CCTTGGGCCATGGAAGGTTGAGTTC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ALOX12P2 PubMed Links

Gene Details

Gene
ALOX12P2
Gene Name
arachidonate 12-lipoxygenase pseudogene 2
There are no transcripts associated with this gene.

View Full Product Details