Product Details

SNP ID
rs4312093
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:1400259 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTCCCGCCCCCTCCATGGCTGGG[A/G]GCTGTTCCCAGGGGCCCTAAGCCTC
Phenotype
MIM: 609236
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BRSK2 PubMed Links

Gene Details

Gene
BRSK2
Gene Name
BR serine/threonine kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256627.1 Intron NP_001243556.1
NM_001256629.1 Intron NP_001243558.1
NM_001256630.1 Intron NP_001243559.1
NM_001282218.1 Intron NP_001269147.1
NM_003957.3 Intron NP_003948.2
XM_005253218.3 Intron XP_005253275.1
XM_006718374.2 Intron XP_006718437.1
XM_006718376.2 Intron XP_006718439.1
XM_011520461.2 Intron XP_011518763.1
XM_011520462.2 Intron XP_011518764.1
XM_017018532.1 Intron XP_016874021.1
XM_017018533.1 Intron XP_016874022.1
XM_017018534.1 Intron XP_016874023.1
XM_017018535.1 Intron XP_016874024.1
XM_017018536.1 Intron XP_016874025.1
XM_017018537.1 Intron XP_016874026.1
XM_017018538.1 Intron XP_016874027.1

View Full Product Details