Product Details

SNP ID
rs4783568
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:66568129 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATAATACTGATAGGAAAACTTGAC[C/T]GAGTTCAGAAAAAGAAAACGAAGTA
Phenotype
MIM: 616074 MIM: 607884
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CKLF PubMed Links

Gene Details

Gene
CKLF
Gene Name
chemokine-like factor
There are no transcripts associated with this gene.

Gene
CKLF-CMTM1
Gene Name
CKLF-CMTM1 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202509.1 Intron NP_001189438.1
NM_001204098.1 Intron NP_001191027.1
NM_001204099.1 Intron NP_001191028.1
Gene
CMTM1
Gene Name
CKLF like MARVEL transmembrane domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052999.3 Intron NP_443725.3
NM_181268.2 Intron NP_851785.2
NM_181269.2 Intron NP_851786.1
NM_181270.2 Intron NP_851787.1
NM_181271.2 Intron NP_851788.1
NM_181272.2 Intron NP_851789.1
NM_181283.2 Intron NP_851800.1
NM_181296.2 Intron NP_851813.1

View Full Product Details