Product Details

SNP ID
rs4334414
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10720997 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGCTGTAAAGTGAGGAGTTAGGTG[C/T]TCTGGGTTGGGAACCTGGGTCTCTC
Phenotype
MIM: 602378 MIM: 609615
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNM2 PubMed Links

Gene Details

Gene
DNM2
Gene Name
dynamin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005360.2 Intron NP_001005360.1
NM_001005361.2 Intron NP_001005361.1
NM_001005362.2 Intron NP_001005362.1
NM_001190716.1 Intron NP_001177645.1
NM_004945.3 Intron NP_004936.2
Gene
MIR638
Gene Name
microRNA 638
There are no transcripts associated with this gene.

Gene
QTRT1
Gene Name
queuine tRNA-ribosyltransferase catalytic subunit 1
There are no transcripts associated with this gene.

View Full Product Details