Product Details

SNP ID
rs4948940
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:45002664 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAACCTACCACTCCCTGTAGAGAA[C/G]TTTCAATTTTCAAAACACTTTATGC
Phenotype
MIM: 610559 MIM: 194529
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C10orf25 PubMed Links

Gene Details

Gene
C10orf25
Gene Name
chromosome 10 open reading frame 25
There are no transcripts associated with this gene.

Gene
RASSF4
Gene Name
Ras association domain family member 4
There are no transcripts associated with this gene.

Gene
ZNF22
Gene Name
zinc finger protein 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006963.4 Intron NP_008894.2

View Full Product Details