Product Details

SNP ID
rs4278407
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:202147723 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAACAGCCAAGCTCTCTAGGTGT[A/T]CTCAGCAAATGCAAGGTATAGTGGG
Phenotype
MIM: 616597 MIM: 176889
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ARL8A PubMed Links

Gene Details

Gene
ARL8A
Gene Name
ADP ribosylation factor like GTPase 8A
There are no transcripts associated with this gene.

Gene
PTPN7
Gene Name
protein tyrosine phosphatase, non-receptor type 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199797.1 2555 UTR 3 NP_001186726.1
NM_002832.3 2555 UTR 3 NP_002823.3
NM_080588.2 2555 UTR 3 NP_542155.1

View Full Product Details