Product Details

SNP ID
rs4407706
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:33392117 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCATGTCTACCGGGAGAGCGAAGG[C/T]CCGTGCGCCCCCGCACAAGTGGAAA
Phenotype
MIM: 603763
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIFC1 PubMed Links

Gene Details

Gene
KIFC1
Gene Name
kinesin family member C1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002263.3 Intron NP_002254.2

View Full Product Details