Product Details
- SNP ID
-
rs4917887
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:100190351 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATTTTTCTTAATGAAAGAGGAAAT[A/G]TACAGAATTGACATTAAGACTAAAT
- Phenotype
-
MIM: 600664
MIM: 611604
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CHUK
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78531297] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CHUK
- Gene Name
- conserved helix-loop-helix ubiquitous kinase
- Gene
- ERLIN1
- Gene Name
- ER lipid raft associated 1
There are no transcripts associated with this gene.
View Full Product Details