Product Details

SNP ID
rs4921442
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:159267092 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACCACCCTCCAGGCCCCAGAATGG[C/G]AGATCTGACAACAGCTTGTACTGTG
Phenotype
MIM: 609867
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
UBLCP1 PubMed Links

Gene Details

Gene
UBLCP1
Gene Name
ubiquitin like domain containing CTD phosphatase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145049.3 Intron NP_659486.2

View Full Product Details