Product Details

SNP ID
rs4790741
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:5086031 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAATTCTCGACAATGGATTCTTACA[A/G]TTGTATAGATACACATTACCTATAA
Phenotype
MIM: 194480
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZFP3 PubMed Links

Gene Details

Gene
ZFP3
Gene Name
ZFP3 zinc finger protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153018.2 Intron NP_694563.1

View Full Product Details