Product Details
- SNP ID
-
rs4942435
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:45781651 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCAGGGGCCCAGCGATTACTGTGTG[A/G]CCAGAGCAGCCTCTGTTATTAAATC
- Phenotype
-
MIM: 615609
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SIAH3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74406725] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SIAH3
- Gene Name
- siah E3 ubiquitin protein ligase family member 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_198849.2 |
2624 |
UTR 3 |
|
|
NP_942146.2 |
View Full Product Details