Product Details

SNP ID
rs4839764
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:99920883 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACATATCGGTACACCTGCCCTTTCT[A/G]ATAATACCAGTAAGATAGACAATCA
Phenotype
MIM: 606111
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MCHR2 PubMed Links

Gene Details

Gene
MCHR2
Gene Name
melanin concentrating hormone receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040179.1 1395 UTR 3 NP_001035269.1
NM_032503.2 1395 UTR 3 NP_115892.2

View Full Product Details