Product Details

SNP ID
rs4364302
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:181064928 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAGAAATGAAGCTTCAAGTTATC[C/T]TTCTAATCCTCCTACCTGGGATTAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00290 PubMed Links

Gene Details

Gene
LINC00290
Gene Name
long intergenic non-protein coding RNA 290
There are no transcripts associated with this gene.

View Full Product Details