Product Details

SNP ID
rs4499546
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:123284111 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCCACAGTCCCTCCAAAGCCAGC[A/T]CTCCTGTGCCAAAGCCCCTGCAGGT
Phenotype
MIM: 600293 MIM: 612442
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ADCY5 PubMed Links

Gene Details

Gene
ADCY5
Gene Name
adenylate cyclase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199642.1 4283 UTR 3 NP_001186571.1
NM_183357.2 4283 UTR 3 NP_899200.1
Gene
SEC22A
Gene Name
SEC22 homolog A, vesicle trafficking protein
There are no transcripts associated with this gene.

View Full Product Details