Product Details
- SNP ID
-
rs4849142
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:113022298 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTTTCTTCCATAGGACTTAACCCCT[A/G]TACATCAGGAAAAGAAATAACAGAT
- Phenotype
-
MIM: 605508
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
IL36B
PubMed Links
Gene Details
- Gene
- IL36B
- Gene Name
- interleukin 36, beta
View Full Product Details