Product Details

SNP ID
rs4802526
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48932988 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATACAGCCAAGAAGGGGCAAGGAG[C/G]GTTTTGAAACCAGTCGGCCAGGCTC
Phenotype
MIM: 606377 MIM: 601323
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DHDH PubMed Links

Gene Details

Gene
DHDH
Gene Name
dihydrodiol dehydrogenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014475.3 198 Intron NP_055290.1
XM_005258748.4 198 UTR 5 XP_005258805.1
XM_017026598.1 198 UTR 5 XP_016882087.1
Gene
NUCB1
Gene Name
nucleobindin 1
There are no transcripts associated with this gene.

View Full Product Details